
Caffeine-13C3 solution

Synonym(s):
Caffeine-trimethyl-13C3 solution
Empirical Formula (Hill Notation):
13C3C5H10N4O2
Molecular Weight:
197.17
EC Number:
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing