
Butalbital solution

Synonym(s):
5-Allyl-5-isobutylbarbituric acid
Empirical Formula (Hill Notation):
C11H16N2O3
CAS Number:
Molecular Weight:
224.26
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing