
Bromoethane-2,2,2-d3

Synonym(s):
Ethyl-2,2,2-d3 bromide
Linear Formula:
CD3CH2Br
CAS Number:
Molecular Weight:
111.98
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing