
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt

Synonym(s):
β-NADPH, 2′-NADPH, Coenzyme II reduced tetrasodium salt, Dihydronicotinamide adenine dinucleotide phosphate tetrasodium salt, NADPH Na4, TPNH2 Na4, Triphosphopyridine nucleotide reduced tetrasodium salt
Empirical Formula (Hill Notation):
C21H26N7Na4O17P3
Molecular Weight:
833.35
EC Number:
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Package Size
Showing 1-4 of 4
Compare
Product Number
Description
Pricing