
Bis(acetonitrile)dichloropalladium(II)

Synonym(s):
Palladium(II) chloride diacetonitrile complex
Linear Formula:
PdCl2 · (CH3CN)2
Products
Product Category
Sustainability
Feature
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing