
Betamethasone acetate

Synonym(s):
Betamethasone 21-acetate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 21-acetate
Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3