Skip to Content
BD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Azlon Square Shoulder Unvented Wash Bottles
Products
Product Category
bottles (16)
Manufacturer
Capacity (mL)
Bottle material
Color
Vial Bottle Material
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
DWKWGW236P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, white polypropylene closure, capacity 500 mL
Expand
View Pricing
DWKWGW443P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, white polypropylene closure, capacity 1000 mL
Expand
View Pricing
DWKWGW237P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, red polypropylene closure, capacity 500 mL
Expand
View Pricing
DWKWGW233P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, green polypropylene closure, capacity 250 mL
Expand
View Pricing
DWKWGW446P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, blue polypropylene closure, capacity 1000 mL
Expand
View Pricing
DWKWGW447P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, green polypropylene closure, capacity 500 mL
Expand
View Pricing
DWKWGW229P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, polypropylene closure (one of each color), capacity 250 mL
Expand
View Pricing
DWKWGW234P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, blue polypropylene closure, capacity 250 mL
Expand
View Pricing
DWKWGW231P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, red polypropylene closure, capacity 250 mL
Expand
View Pricing
DWKWGW239P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, green polypropylene closure, capacity 500 mL
Expand
View Pricing
DWKWGW445P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, red polypropylene closure, capacity 1000 mL
Expand
View Pricing
DWKWGW230P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, white polypropylene closure, capacity 250 mL
Expand
View Pricing
DWKWGW232P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, yellow polypropylene closure, capacity 250 mL
Expand
View Pricing
DWKWGW235P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, blue polypropylene closure, capacity 500 mL
Expand
View Pricing
DWKWGW238P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, yellow polypropylene closure, capacity 500 mL
Expand
View Pricing
DWKWGW444P
Azlon Square Shoulder Unvented Wash Bottles,
wide-neck, low-density polyethylene bottle, yellow polypropylene closure, capacity 1000 mL
Expand
View Pricing