
Atovaquone for system suitability

Synonym(s):
Atovaquone, Mepron, trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione
Empirical Formula (Hill Notation):
C22H19ClO3
CAS Number:
Molecular Weight:
366.84
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing