
Atomoxetine hydrochloride

Synonym(s):
(R)-Tomoxetine hydrochloride, (R)-N-Methyl-γ-(2-methylphenoxy)benzenepropanamine hydrochloride, Atomoxetine hydrochloride
Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing