
Asp-Phe

Synonym(s):
L-Aspartyl-L-phenylalanine, Aspartame acid
Linear Formula:
HOOCCH2CH(NH2)CONHCH(CH2C6H5)CO2H
CAS Number:
Molecular Weight:
280.28
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Reaction Type
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2