
Aspalathin

Synonym(s):
3-(3,4-Dihydroxyphenyl)-1-(3-β-D-glucopyranosyl-2,4,6-trihydroxyphenyl)-1-propanone, 3-(3,4-Dihydroxyphenyl)-3′-β-D-glucopyranosyl-2′,4′,6′-trihydroxypropiophenone
Empirical Formula (Hill Notation):
C21H24O11
CAS Number:
Molecular Weight:
452.41
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing