
Artemisinin

Synonym(s):
Arteannuin, Qinghaosu
Empirical Formula (Hill Notation):
C15H22O5
CAS Number:
Molecular Weight:
282.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4