
Arbutin

Synonym(s):
4-Hydroxyphenyl-β-D-glucopyranoside, p-Arbutin, Hydroquinone β-D-glucopyranoside
Empirical Formula (Hill Notation):
C12H16O7
CAS Number:
Molecular Weight:
272.25
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4