
Acibenzolar acid

Synonym(s):
1,2,3-Benzothiadiazole-7-carboxylic acid
Empirical Formula (Hill Notation):
C7H4N2O2S
CAS Number:
Molecular Weight:
180.18
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing