
8-Hydroxyquinoline copper(II) salt

Linear Formula:
Cu(C9H6NO)2
CAS Number:
Molecular Weight:
351.85
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing