Skip to Content
Merck
5-Chloro-1-methyl-4-nitroimidazole British Pharmacopoeia (BP) Reference Standard

5-Chloro-1-methyl-4-nitroimidazole

5-Chloro-1-methyl-4-nitroimidazole British Pharmacopoeia (BP) Reference Standard
Empirical Formula (Hill Notation):
C4H4ClN3O2
CAS Number:
Molecular Weight:
161.55

Products

Product Category
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2