Skip to Content
Merck
4-Hydroxybenzenesulfonic acid solution 65 wt. % in H2O

4-Hydroxybenzenesulfonic acid solution

4-Hydroxybenzenesulfonic acid solution 65 wt. % in H2O
Synonym(s):
Phenol-4-sulfonic acid
Linear Formula:
HOC6H4SO3H
CAS Number:
Molecular Weight:
174.17

Products

Product Category
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing