Skip to Content
Merck
4-Formylbenzene-1,3-disulfonic acid disodium salt hydrate 97%

4-Formylbenzene-1,3-disulfonic acid disodium salt hydrate

4-Formylbenzene-1,3-disulfonic acid disodium salt hydrate 97%
Synonym(s):
Benzaldehyde 2,4-disulfonic acid disodium salt, Sodium 4-formylbenzene-1,3-disulfonate
Linear Formula:
HCOC6H3(SO3Na)2 · xH2O
CAS Number:
Molecular Weight:
310.21 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing