
4,5-Dihydroxy-1,3-benzenedisulfonic acid disodium salt monohydrate

Linear Formula:
(OH)2C6H2(SO3Na)2 · H2O
Products
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing