Skip to Content
Merck
4-(3,6-Dimethyl-3-heptyl)phenol-diethoxylate-ring-13C6 solution 10 μg/mL in acetone, analytical standard

4-(3,6-Dimethyl-3-heptyl)phenol-diethoxylate-ring-13C6 solution

4-(3,6-Dimethyl-3-heptyl)phenol-diethoxylate-ring-13C6 solution 10 μg/mL in acetone, analytical standard
Synonym(s):
2-{2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy]ethoxy}ethanol-13C6, 2-{2-[4-(3,6-Dimethyl-3-heptyl)phenoxy]ethoxy}ethanol-13C6, 3,6,3-Nonylphenol diethoxylate-13C6, 363-NP2EO-13C6
Empirical Formula (Hill Notation):
13C6C13H32O3
CAS Number:
Molecular Weight:
314.41
EC Number:

Products

Brand
Formula Weight
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing