
2-Oxo Clopidogrel (Mixture Of Diastereomers)

Synonym(s):
(methyl 2-(2-chlorophenyl)-2-(2-oxo-2,6,7,7a-tetrahydrothieno[3,2-c]pyridin-5(4H)-yl)acetate)
Empirical Formula (Hill Notation):
C16H16ClNO3S
CAS Number:
Molecular Weight:
337.82
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing