![2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline ≥99% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/308/719/159746c3-f2b8-48bf-8ca1-a70a160b4a73/640/159746c3-f2b8-48bf-8ca1-a70a160b4a73.png)
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline
![2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline ≥99% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/308/719/159746c3-f2b8-48bf-8ca1-a70a160b4a73/640/159746c3-f2b8-48bf-8ca1-a70a160b4a73.png)
Synonym(s):
Bathocuproine, 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, BCP
Empirical Formula (Hill Notation):
C26H20N2
CAS Number:
Molecular Weight:
360.45
Products
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing