
2,4-Dichlorophenylacetic acid

Linear Formula:
Cl2C6H3CH2CO2H
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing