Skip to Content
Merck
2,3-Dichloroquinoxaline 96%

2,3-Dichloroquinoxaline

2,3-Dichloroquinoxaline 96%
Empirical Formula (Hill Notation):
C8H4Cl2N2
CAS Number:
Molecular Weight:
199.04
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing