
2-(2-Bromoethyl)-1,3-dioxolane

Synonym(s):
3-Bromopropionaldehyde ethylene acetal
Empirical Formula (Hill Notation):
C5H9BrO2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1