
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, polymer-bound

Synonym(s):
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide polymer-bound, N-Ethyl-N′-(3-dimethylaminopropyl)carbodiimide polymer-bound, EDC polymer-bound
EC Number:
Products
Product Category
Brand
Physical Form
Chain Type
Reaction Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing