Merck
HomeProductsLab Water Purification SystemsMilli-Q® Benchtop Lab Water Purification Systems
Milli-Q® Benchtop Lab Water Purification Systems
Milli-Q® IQ 7003/05/10/15 Ultrapure and Pure Water Purification System - Type 1 and Type 2 lab water purification system with production and tank units plus pure and ultrapure water dispensers.
Your Ultimate Lab Water Experience: Milli-Q® IQ 7003/05/10/15 Ultrapure and Pure Water Purification System
The Milli-Q® Lab Water Solutions portfolio of benchtop water systems includes both pure and ultrapure systems to meet any lab’s needs. From sensitive QC analyses to buffer preparation and glassware rinsing, this collection of compact systems has your lab covered. Our specialists can help you find the water purification solution that’s best adapted to your requirements in terms of:   
Products
Sort by Relevance
Showing 1-20 of 40
Change View
Showing 1-20 of 40
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 40 results
Page 1 of 2
Page 1 of 2
What are the different lab water grades?
Water grades in the lab are defined by quality standards, such as the ASTM or ISO 3696, which help ensure that the correct water quality is used for specific applications. Standards also consider the cost of production, as Type 1 ultrapure water is more expensive to produce than Type 2 or Type 3 pure water.
Type 1 ultrapure water is ion-free (resistivity 18.2 MΩ·cm) with low total organic carbon (TOC) content. Ultrapure water is recommended for sensitive laboratory applications such as advanced analytical techniques (e.g. HPLC, UHPLC, LC-MS, ICP-MS, GF-AAS), molecular biology applications (e.g. PCR, sequencing), and to prepare buffers and culture media for mammalian cell culture.
Type 2 pure water is recommended for general lab use, such as to prepare buffers, reagents, and culture media, and as feed for other equipment (e.g. Type 1 systems, clinical analyzers, dishwashers, stability test chambers, etc.). Our Type 2 systems incorporate environmentally friendly Elix® electrodeionization (EDI) technology which produces constant-quality pure water while eliminating the need for hazardous chemical regeneration procedures, costly resins, changing DI cartridges, and adding softeners.
Type 3 water is the lowest water grade and is produced by reverse osmosis (RO). RO water is recommended for non-critical lab applications, such as glassware rinsing, heating baths, autoclaves, and as feed for Type 1 water systems.
Clinical Laboratory Reagent Water (CLRW) complies with quality standards described by the Clinical and Laboratory Standards Institute (CLSI®) guideline, which defines water purity in clinical laboratories (resistivity > 10 MΩ·cm; TOC < 500 ppb; bacteria < 10 cfu/mL; filtration of particles > 0.22 μm).
Scientist dispensing ultrapure water from Q-POD® dispenser
Space-saving and Flexible Options—From Installation to Dispensing
Our benchtop systems are designed to save valuable lab space, providing versatile installation, storage, and dispensing options. Our most recent Milli-Q® IQ, IX and EQ series systems require only the slim POD dispenser on the bench or mounted on the wall. The compact purification unit and tank can be integrated under the bench, on the wall, or a combination of both to suit your requirements.
For labs needing more than one point-of-use, multiple dispensers can be connected to one Milli-Q® IQ or Milli-Q® IX system, with up to 5 m between the PODs. This way, scientists at distant benches—or even another lab—have access to purified water. In addition, a range of production flow rates and tank size options are available to ensure your lab's needs are fully met.
A touchscreen on the Q-POD® dispenser eases use and water quality monitoring
Digital Services Simplify Water System Care and assure Productivity
The MyMilli-Q™ platform is a unique online solution that simplifies the care of your Milli-Q® water system and can support maximum lab productivity. Here are some of the tasks you can accomplish by simply logging into your MyMilli-Q™ account:
  • Track service history
  • Download service reports
  • Schedule maintenance visits
  • Track consumable orders
  • Manage consumable deliveries
  • View contact details
  • Renew your service plan
  • Contact Milli-Q® Services
With these time-saving features, you’ll benefit from:
  • At-a-glance, consolidated contract information
  • More rapid and efficient planning
  • Streamlined audit preparation
  • Easily traceable service history and reports
MyMilli-Q™ Digital Services Sticker Icon
Plus, with a subscription to MyMilli-Q™ Remote Care online monitoring and service feature (available on Milli-Q® IQ and Milli-Q® IX series systems), you’ll benefit from:
  • Online access to your system and its data
  • Automatic notifications, by email or SMS, in case of an alert or alarm
  • Faster service with remote diagnostics and repair possible
  • Simplified data management and advanced traceability for eased audit and accreditation processes
MyMilli-Q™ Digital Services are available for all systems under warranty or covered by a Milli-Q® Service Plan.
Greener Water Systems
To reduce the environmental impact of water purification in the lab, Milli-Q® water systems and processes are becoming increasingly more sustainable. Here are some ways that newer generation Milli-Q® water systems can support your lab to achieve greater sustainability:
  • Water-saving purification technologies
  • Energy-saving design and system features
  • Reduced chemical waste
  • Mercury-free UV lamps
  • Greener packaging
  • Paperless data management
  • Remote diagnostics and services
Learn how Milli-Q® water systems can help your lab to go greener.
Related Resources
  • Milli-Q® IQ 7003/05/10/15 Ultrapure & Pure Lab Water Purification System
    Milli-Q® IQ 7003/05/10/15 lab water systems provide consistent-quality Type 1 and Type 2 water, are intuitive to use, and can support your lab's sustainability goals.
  • Milli-Q® IQ 7000 Ultrapure Water System
    Discover an ultrapure lab water system designed with you in mind. Intelligent, intuitive and connected, the Milli-Q® IQ 7000 system will simplify the way you work in the lab.
  • Milli-Q® IX Pure Lab Water System
    Discover Milli-Q® IX systems for high-purity water. Ideal for general lab & sensitive techniques. Reliable Efficient Innovative. A Greener Alternative.
  • EN ISO 11133 Microbiological Culture Media
    Water source comparison for microbiological media preparation highlights central purification vs. Milli-Q® IX system efficiency.
  • Brochure: Milli-Q® EQ 7008/16 Ultrapure & Pure (RO) Water System
    Milli-Q® EQ 7008/7016 ultrapure and reverse osmosis (RO) water systems are designed to produce consistent ultrapure water quality [resistivity 18.2 MΩ·cm @ 25 °C; total organic carbon (TOC) ≤ 5 ppb] direct from a tap water source. Final ultrapure water quality can be adapted to each user’s specific application requirements. Plus, a range of intelligent design features make it effortless to dispense water as needed and where needed in the lab.
  • Brochure: Milli-Q® EQ 7000 Ultrapure Water System
    The Milli-Q® EQ 7000 ultrapure water system is designed to produce consistent ultrapure water quality [resistivity 18.2 MΩ·cm @ 25 °C; total organic carbon (TOC) ≤ 5 ppb] that can be adapted to each user’s specific application requirements. Plus, a range of intelligent design features make it effortless to dispense water as needed and where needed in the lab.
  • Brochure: Milli-Q® IQ Element for Trace Elemental Analysis
    The Milli-Q® IQ Element unit, combined with a Milli-Q® IQ 7 series water purification system, delivers analytical-grade ultrapure water that is suitable for trace and ultra-trace elemental analyses, including ICP-MS, GF-AAS and trace IC.
  • Flyer: Online Contract Management
    Milli-Q® system contract management is now only a click away! Online Contract Management is our digital service that streamlines the care of your Milli-Q® fleet so that you can better focus on your job priorities.
  • Sustainability Milli-Q® Lab Water Purification Systems
    Reduce your lab’s consumption of water, energy, plastics and chemicals. Eco-friendly Milli-Q® lab water purification systems and digital services can support your lab’s sustainability goals.
Page 1 of 2
Related Videos