Skip to Content
Merck
All Photos(1)

Key Documents

21929-U

SGE Syringes for PerkinElmer® Autosystem

volume 5 μL, 5F-PE-GT-0.63, needle size 23 ga, needle L 70 mm, needle type, LTN (fixed)

Synonym(s):

TM=["PerkinElmer"] Autosystem Syringe by SGE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

cone tip stainless steel needle (point style 1)

needle size

23 ga

description

color (lime)

feature

needle type LTN (fixed)
gastight syringe, fixed needle

packaging

pkg of 1 ea

manufacturer/tradename

SGE 001957

needle L

70 mm

volume

5 μL

Looking for similar products? Visit Product Comparison Guide

General description

SGE syringes [volume 5 μL, 5F-PE-GT-0.63, needle size 23 ga (cone tip), needle L 70 mm, needle type, LTN (fixed)] are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most trouble-free autosampler syringes available. The stainless steel cone tip, point style #1 is recommended for use with pre-drilled septa. The shape of this needle on the syringe has been developed for multiple and fast septum injections on the autosampler.

Legal Information

PerkinElmer is a registered trademark of PerkinElmer Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service