Skip to Content
Merck
All Photos(8)

Key Documents

ZMS1195

Sigma-Aldrich

Anti-PVR/CD155 Antibody, clone SKII.4 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CD155, NECL-5, Nectin-like protein 5, Poliovirus receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

SKII.4, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 45.3 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone SKII.4 is ZooMAb® mouse recombinant monoclonal antibody that specifically detects Poliovirus receptor (PVR/CD155). It targets an epitope within the extracellular domain.

Immunogen

SK-N-S1 human neuroblastoma cells.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected PVR/CD155 in HeLa cells.

Tested Applications

Flow Cytometry Analysis: 0.1 µg from a representative lot detected PVR/CD155 in one million Hela cells.

Affinity Binding Assay: A representative lot of this antibody bound His-tagged recombinant fragment corresponding to the extracellular domain of Human PVR/CD155 with a KD of 1.4 x 10-6 in an affinity binding assay.

Enzyme Immunoassay Analysis (ELISA): A serial of dilutions from a representative lot detected His-tagged recombinant fragment corresponding to the extracellular domain of Human PVR/CD155.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Poliovirus receptor (UniProt: P15151; also known as Nectin-like protein 5, NECL-5, CD155) is encoded by the PVR (also known as PVS) gene (Gene ID: 5817) in human. CD155 is a single-pass type I, N-glycosylated, membrane protein that mediates NK cell adhesion and triggers NK cell effector functions. It is ubiquitously expressed, and many human tumors are reported to significantly upregulate its expression. It binds two different NK cell receptors: CD96 and CD226 and these interactions accumulate at the cell-cell contact site, leading to the formation of a mature immunological synapse between NK cell and the target cell. This may trigger adhesion and secretion of lytic granules and IFN-γ and activate cytotoxicity of activated NK cells. CD155 serves as a receptor for poliovirus and may play a role in axonal transport of poliovirus by targeting virion-PVR-containing endocytic vesicles to the microtubular network through interaction with dynein light chain Tctex-type 1 (DYNLT1). It is synthesized with a signal peptide (aa 1-20), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 21-343), a transmembrane domain (aa 344-367), and a cytoplasmic domain (aa 368-417). It contains two Ig-like C2 type domains (aa 145-237 and 233-328), and an Ig-like V-type domain that is shown to be necessary and sufficient for virus binding and uptake. CD155 also has one the immunoreceptor tyrosine-based inhibitor motif (ITIM; aa 396-401) that when phosphorylated by Src kinases binds to the SH2 domain of several SH2-containing phosphatases. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sloan, KE., et al. (2004). BMC Cancer. 4; 73; Oda, T., et al. (2004). Biochem. Biophys. Res. Commun. 319(4);1253-1264; Mendelsohn, CL., et al. (1989). Cell. 56(5); 855-865).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service