Z742383
CellCrown™ inserts
12 well plate inserts, sterile
Synonym(s):
3D cell inserts
All Photos(1)
About This Item
Recommended Products
material
clear polycarbonate
polycarbonate
sterility
sterile; γ-irradiated
sterile
feature
lid
packaging
pack of 6 ea
manufacturer/tradename
Scaffdex C00007S
size
12 well
General description
CellCrown is a disposable insert, which immobilizes a sample in its place on a 6, 12, 24, 48, and 96-well plate. Fibers, foils, membranes, textiles, tissue and block like samples can be fixed with CellCrown in a firm, adjusted position in the well. The sample can be left in the well during the change of media by means of a pipette between the wall of the well and the CellCrown. The lid of the well plate can be closed while CellCrown is used. The insert can be removed from the well plate and unloaded after cell culturing without damaging the sample. CellCrown is made of medical grade polycarbonate and the product range includes sterile products ready to use as well as non-sterile products to be sterilized by the user. Sterilization can be by gamma or e-beam radiation, autoclaving, ethylene oxide gas and plasma sterilization.
Sterilization Conditions:
Temperature.......121 °C / 250 °F
Time..................15 min.
Drying time.........15 min.
Sterilization Conditions:
Temperature.......121 °C / 250 °F
Time..................15 min.
Drying time.........15 min.
Legal Information
CellCrown is a trademark of Scaffdex Oy