Skip to Content
Merck
All Photos(2)

Key Documents

SML3909

Sigma-Aldrich

SGC-CK2-2

≥98% (HPLC)

Synonym(s):

5-(Benzylamino)pyrimido[4,5-c]quinoline-8-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14N4O2
CAS Number:
Molecular Weight:
330.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

SGC-CK2-2 is a highly potent and selective casein kinase 2 (CK2, CKII) inhibitor (IC50 = 3 nM/CK2α and <1 nM/CK2α by enzyme assay; CK2α/CK2α′ IC50 = 920/200 nM by cellular NanoBRET; genomewide by DiscoverX scanMAX panel at 1 μM). SGC-CK2-2 effectively inhibits CK2 downstream signaling in MDA-MB-231 cells (AKT pS129 IC50 ~5 μM post 24h) without cytotoxicity (up to 10 μM & 48h).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of a Potent and Selective Naphthyridine-Based Chemical Probe for Casein Kinase 2
ACS Medicinal Chemistry Letters, 14(4), 432-441 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service