Skip to Content
Merck
All Photos(1)

Key Documents

SML1813

Sigma-Aldrich

Ani9

Synonym(s):

(4-Chloro-2-methylphenoxy)-acetic acid [(2-methoxyphenyl)methylene]hydrazide, 2-(4-Chloro-2-methylphenoxy)-N-[(2-methoxyphenyl)methylideneamino]-acetamide, 2-(4-Chloro-2-methylphenoxy)-acetic acid 2-[(2-methoxyphenyl)methylene]hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClN2O3
CAS Number:
Molecular Weight:
332.78
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(OCC(NN=CC2=C(OC)C=CC=C2)=O)C=CC(Cl)=C1

InChI key

KDALDZRKOBJXIE-UHFFFAOYSA-N

Application

Ani9 has been used to find out the inappropriate modulation of the membrane protein functions that are involved in the 198 ADGRG2- or Gq-mediated regulation of fluid reabsorption in the efferent ductules.[1]

Biochem/physiol Actions

2-(4-chloro-2-methylphenoxy)-N-[(2-methoxyphenyl)methylideneamino]-acetamide (Ani9), the most potent inhibitor, is the structural analog of Ani7.[2] It may be considered as a novel candidate for drug therapy of cancer, hypertension, pain, diarrhea and asthma.[2]
Ani9 is a potent and highly selective anoctamin1 (ANO1)/transmembrane protein 16A (TMEM16A) inhibitor that completely inhibited ANO1 chloride current with submicromolar potency. Ani9 does not affect the intracellular calcium signaling and CFTR chloride channel activity.
Ani9 is a potent and highly selective anoctamin1 (ANO1)/transmembrane protein 16A (TMEM16A) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gq activity-and β-arrestin-1 scaffolding-mediated ADGRG2/CFTR coupling are required for male fertility.
Zhang DL, et al.
eLife, 7, e33432-e33432 (2018)
Ani9, a novel potent small-molecule ANO1 inhibitor with negligible effect on ANO2.
Seo Y, et al.
PLoS ONE, 11(5), e0155771-e0155771 (2016)
Alexander Klimovich et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(30), 17854-17863 (2020-07-11)
Pacemaker neurons exert control over neuronal circuit function by their intrinsic ability to generate rhythmic bursts of action potential. Recent work has identified rhythmic gut contractions in human, mice, and hydra to be dependent on both neurons and the resident

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service