Skip to Content
Merck
All Photos(3)

Key Documents

HPA021676

Sigma-Aldrich

Anti-RACK1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cell proliferation-inducing gene 21 protein, Anti-Guanine nucleotide-binding protein subunit beta-2-like 1, Anti-Guanine nucleotide-binding protein subunit beta-like protein 12.3, Anti-HLC-7, Anti-RACK1, Anti-Receptor for activated C kinase, Anti-Receptor of activated protein kinase C 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

mouse, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 0.04-0.4 μg/mL

immunogen sequence

CVRFSPNSSNPIIVSCGWDKLVKVWNLANCKLKTNHIGHTGYLNTVTVSPDGSLCASGGKDGQAMLWDLNEGKHLYTLDG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GNB2L1(10399)

General description

Guanine nucleotide binding protein (G protein), β polypeptide 2-like 1 (GNB2L1) is a stably expressed housekeeping gene localized in alveolar macrophage and neutrophils. The gene has been predicted to encode a protein RACK1 (Receptor for activated C kinase 1). It is mapped on the telomeric position of chromosome 5q35.3.

Immunogen

Guanine nucleotide-binding protein subunit beta-2-like 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-GNB2L1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Guanine nucleotide binding protein (G protein), β polypeptide 2-like 1 (GNB2L1) is a scaffold protein highly involved in the binding and anchorage of protein kinase C. It has been predicted that GNB2L1 may act as a regulatory cofactor of multidrug resistance protein (MDR3/ABCB4) and is vital for the plasma membrane localization and translocation function of ABCB4.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74729

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Ikebuchi et al.
Hepatology research : the official journal of the Japan Society of Hepatology, 39(11), 1091-1107 (2009-08-14)
Multidrug resistance protein 3 (MDR3/ABCB4), located on the bile canalicular membrane of hepatocytes, is responsible for the translocation of phosphatidylcholine across the plasma membrane, and its hereditary defect causes liver disorders, such as progressive familial intrahepatic cholestasis type 3. We
Xiaozhu Zhang et al.
BMC molecular biology, 6, 4-4 (2005-02-22)
Reference genes, which are often referred to housekeeping genes, are frequently used to normalize mRNA levels between different samples. However the expression level of these genes may vary among tissues or cells, and may change under certain circumstances. Thus the
Motoyuki Otsuka et al.
PloS one, 6(9), e24359-e24359 (2011-09-22)
MicroRNAs (miRNAs) are important regulators of gene expression that control physiological and pathological processes. A global reduction in miRNA abundance and function is a general trait of human cancers, playing a causal role in the transformed phenotype. Here, we sought
Shu Wang et al.
Molecular biology reports, 30(1), 53-60 (2003-04-12)
During a large-scale screen of a human fetal brain cDNA library, a novel human gene GNB2L1 encoding a novel RACK (receptor of activated protein kinase C) protein was isolated and sequenced. The cDNA is 1142 bp long and has a
T Ishii et al.
The European respiratory journal, 27(2), 300-306 (2006-02-03)
The stability of housekeeping genes is critical when performing gene expression studies. To date, there have been no studies that look at the stability of commonly used housekeeping genes in alveolar macrophages. Expression levels may be affected by culture, stimulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service