Skip to Content
Merck
All Photos(4)

Key Documents

HPA018679

Sigma-Aldrich

Anti-RASGRF2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Guanine nucleotide-releasing factor 2, Anti-Ras guanine nucleotide exchange factor 2, Anti-Ras-GRF2, Anti-Ras-specific guanine nucleotide-releasing factor 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

FATSQNNRGEHLVDGKSPRLCRKFSSPPPLAVSRTSSPVRARKLSLTSPLNSKIGALDLTTSSSPTTTTQSPAASPPPHTGQIPLDLSRGLSSPEQSPGTVEENVD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RASGRF2(5924)

General description

The gene Ras-specific guanine nucleotide-releasing factor-2 (RASGRF2) is mapped to human chromosome 5q13. It belongs to a family of calcium/calmodulin-regulated guanine-nucleotide exchange factors. RASGRF2 transcripts are abundantly expressed in human brain tissue. Low levels of RASGRF2 are also detected in human heart, placenta, kidney, pancreas, human ovary and spleen tissues. The protein localizes to the cytoplasm. However, depending upon the interaction partner it can translocate to the cell periphery. RasGRF2 protein contains two pleckstrin homology (PH) regions, a coiled-coil motif, a Ca2+/calmodulin binding ilimaquinone (IQ) domain, a Dbl homology (DH) region and the prototypical Cdc25 (Ras-specific guanine nucleotide-releasing factor 1) Ras exchange domain.

Immunogen

Ras-specific guanine nucleotide-releasing factor 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In neurons, p35/CDK5 (cyclin-dependent like kinase 5) phosphorylates Ras-specific guanine nucleotide-releasing factor-2 (RASGRF2), thereby modulating Rac-dependent extracellular signal-regulated kinase (ERK1/2) activity and microtubule-associated protein-1b distribution. RASGRF2 is also involved in T-cell signaling. Presence of RASGRF2 in T-cells activates Ras and stimulates the transcriptional factor NF-AT (nuclear factor of activated T cells).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74632

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sashi Kesavapany et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 24(18), 4421-4431 (2004-05-07)
Cyclin-dependent kinase 5 (Cdk5) is a proline-directed kinase the activity of which is dependent on association with its neuron-specific activators, p35 and p39. Cdk5 activity is critical for the proper formation of cortical structures and lamination during development. In the
H Shuen Lo et al.
Genome research, 13(8), 1855-1862 (2003-08-07)
Variations in gene sequence and expression underlie much of human variability. Despite the known biological roles of differential allelic gene expression resulting from X-chromosome inactivation and genomic imprinting, a large-scale analysis of allelic gene expression in human is lacking. We
Sergio Ruiz et al.
Molecular and cellular biology, 27(23), 8127-8142 (2007-10-10)
The Ras pathway is critical for the development and function of T lymphocytes. The stimulation of this GTPase in T cells occurs primarily through the Vav1- and phospholipase C-gamma1-dependent activation of RasGRP1, a diacylglycerol-responsive Ras GDP/GTP exchange factor. Here, we
Mohini Lutchman et al.
European journal of biochemistry, 269(2), 638-649 (2002-02-22)
Erythroid dematin is a major component of red blood cell junctional complexes that link the spectrin-actin cytoskeleton to the overlying plasma membrane. Transcripts of dematin are widely distributed including human brain, heart, lung, skeletal muscle, and kidney. In vitro, dematin
Xabier L Aranguren et al.
Blood, 122(24), 3982-3992 (2013-10-11)
Endothelial cells (ECs) lining arteries and veins have distinct molecular/functional signatures. The underlying regulatory mechanisms are incompletely understood. Here, we established a specific fingerprint of freshly isolated arterial and venous ECs from human umbilical cord comprising 64 arterial and 12

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service