Skip to Content
Merck
All Photos(1)

Key Documents

TR6Bc1

85042301, mouse nerve, Glial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

TR6Bc1, 85042301

biological source

mouse nerve

growth mode

Adherent

karyotype

Not specified

morphology

Glial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C3H.He Schwannoma

Cell Line Description

Ethyl nitrosourea induced tumour in C3H. The mice, originated in trigeminal nerve and brain; readily transplanted and resembles schwannoma histologically. Cultured cells form prominent processes, resemble 33B (Sigma Catalogue number. 85081901) morphologically, contain abundant C type particles.

Application

Tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 10% Tryptose Phosphate Broth + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service