Skip to Content
Merck
All Photos(1)

Key Documents

B2938

Sigma-Aldrich

BEPP monohydrochloride

≥98% (HPLC)

Synonym(s):

1H-Benzimidazole-1-ethanol, 2,3-dihydro-2-imino-alpha-(phenoxymethyl)-3-(phenylmethyl)- monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23N3O2 HCl
CAS Number:
Molecular Weight:
409.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Cl.OC(COc1ccccc1)CN2C(=N)N(Cc3ccccc3)c4ccccc24

InChI

1S/C23H23N3O2.ClH/c24-23-25(15-18-9-3-1-4-10-18)21-13-7-8-14-22(21)26(23)16-19(27)17-28-20-11-5-2-6-12-20;/h1-14,19,24,27H,15-17H2;1H

InChI key

KDMXJFUGIDGZQL-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

BEPP is a double-strand RNA-dependent protein kinase (PKR) activator.
BEPP is a double-strand RNA-dependent protein kinase (PKR) activator. The double-strand RNA (dsRNA)-dependent protein kinase is a ubiquitously expressed serine threonine kinase, inducible by interferon γ. PRK is involved in several curtail cellular regulations including phosphorylation of eIF2α (which leads to inhibition of protein synthesis and eliciting antivirus and antitumor activities), modulating activities of eIF2α, NF-κB, ATF-3, and p53. BEPP also inhibits the growth of a human lung cancer cell line overexpressing PKR.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana-Belén Blázquez et al.
PLoS neglected tropical diseases, 15(1), e0009072-e0009072 (2021-01-26)
Usutu virus (USUV) is an African mosquito-borne flavivirus closely related to West Nile, Japanese encephalitis, Zika, and dengue viruses. USUV emerged in 1996 in Europe, where quickly spread across the continent causing a considerable number of bird deaths and varied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service