Skip to Content
Merck
All Photos(2)

Key Documents

11813

Sigma-Aldrich

Potassium cyanide

technical, ≥96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCN
CAS Number:
Molecular Weight:
65.12
Beilstein:
3593645
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
SB.54

grade

technical

Assay

≥96%

form

solid

pH

11.5 (20 °C, 20 g/L)

mp

634 °C (lit.)

cation traces

Na: ≤5000 mg/kg

SMILES string

[K]C#N

InChI

1S/CN.K/c1-2;

InChI key

YUZRZFQHUCKACF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Characterization, antimicrobial and antitumor activity of superoxide dismutase extracted from Egyptian honeybee venom (Apis mellifera lamarckii).: This study explores the extraction and characterization of superoxide dismutase from honeybee venom, highlighting its antimicrobial and antitumor properties. Potassium cyanide was used in the biochemical analysis to understand the enzyme′s activity and stability (Abdel-Monsef et al., 2023).

  • Cytochromes P450 2C8 and 3A Catalyze the Metabolic Activation of the Tyrosine Kinase Inhibitor Masitinib.: This research investigates the metabolic pathways of masitinib, focusing on the role of cytochromes P450. Potassium cyanide is used to inhibit specific enzymatic activities during the analysis, providing insights into drug metabolism and potential toxicological impacts (Latham et al., 2022).

  • Profiling of in vivo, in vitro and reactive zorifertinib metabolites using liquid chromatography ion trap mass spectrometry.: The study details the metabolic profiling of zorifertinib using advanced chromatographic techniques. Potassium cyanide serves as an analytical reagent to assess the stability and reactivity of various metabolites (Al-Shakliah et al., 2022).

  • Structural and Biochemical Characterization of a Dye-Decolorizing Peroxidase from Dictyostelium discoideum.: The research characterizes a peroxidase enzyme from Dictyostelium discoideum, utilizing potassium cyanide in the enzymatic assays to inhibit specific reactions and better understand the enzyme′s functionality (Rai et al., 2021).

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of the Methiodide of N, N-Dimethylaminomethylferrocene with Potassium Cyanide to Form Ferrocylacetonitrile1.
Lednicer D, et al.
The Journal of Organic Chemistry, 23(5), 653-655 (1958)
Eagleson M.
Concise Encyclopedia Chemistry, 887-887 (1994)
The determination of amino-acids with ninhydrin.
Yemm EW, et al.
Analyst, 80(948), 209-214 (1955)
Catalytic asymmetric synthesis of O-acetylcyanohydrins from potassium cyanide, acetic anhydride, and aldehydes, promoted by chiral salen complexes of titanium (IV) and vanadium (V).
Belokon YN, et al.
Helvetica Chimica Acta, 85(10), 3301-3312 (2002)
P Lawson-Smith et al.
Undersea & hyperbaric medicine : journal of the Undersea and Hyperbaric Medical Society, Inc, 38(4), 223-237 (2011-09-01)
Cyanide (CN) intoxication inhibits cellular oxidative metabolism and may result in brain damage. Hydroxycobalamin (OHCob) is one among other antidotes that may be used following intoxication with CN. Hyperbaric oxygen (HBO2) is recommended when supportive measures or antidotes fail. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service