Skip to Content
Merck
All Photos(1)

Key Documents

04272

Sigma-Aldrich

di-Sodium hydrogen phosphate dihydrate

meets analytical specification of Ph. Eur., BP, 98.5-101% (calc. to the dried substance)

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 2H2O
CAS Number:
Molecular Weight:
177.99
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Agency

meets analytical specifications of BP
meets analytical specifications of Ph. Eur.

Assay

98.5-101% (calc. to the dried substance)

form

crystals

quality

meets analytical specification of Ph. Eur., BP

impurities

reducing matter, complies
residual solvents, complies
≤0.001% heavy metals (as Pb)
≤1% NaH2PO4 (calc. to dried subst.)

loss

19.5-21% loss on drying, 130 °C

pH

9.0-9.4 (20 °C, 1%)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Fe: ≤10 mg/kg

suitability

complies for appearance of solution

SMILES string

O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.2H2O/c;;1-5(2,3)4;;/h;;(H3,1,2,3,4);2*1H2/q2*+1;;;/p-2

InChI key

KDQPSPMLNJTZAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium phosphate dibasic dihydrate has been used for isolating DNA and RNA from bacterial samples of human gastrointestinal (GI) tract.

Biochem/physiol Actions

Sodium phosphate dibasic dihydrate is an important component of running buffer of denaturing gel electrophoresis.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erwin G Zoetendal et al.
Nature protocols, 1(2), 954-959 (2007-04-05)
The human gastrointestinal (GI) tract contains a complex microbial community that consists of numerous uncultured microbes. Therefore, nucleic-acid-based approaches have been introduced to study microbial diversity and activity, and these depend on the proper isolation of DNA, rRNA and mRNA.
Erwin G Zoetendal et al.
Nature protocols, 1(2), 870-873 (2007-04-05)
The human gastrointestinal (GI) tract contains a complex microbial community that develops in time and space. The most widely used approaches to study microbial diversity and activity are all based on the analysis of nucleic acids, DNA, rRNA and mRNA.
Sohini Mukherjee et al.
Nature communications, 6, 8467-8467 (2015-10-13)
Creating an artificial functional mimic of the mitochondrial enzyme cytochrome c oxidase (CcO) has been a long-term goal of the scientific community as such a mimic will not only add to our fundamental understanding of how CcO works but may
Patrizia Iavicoli et al.
Journal of chromatography. A, 1422, 260-269 (2015-10-27)
Asymmetric Flow Field-Flow Fractionation (AF4) combined with multidetector analysis form a promising technique in the field of nanoparticle characterization. This system is able to measure the dimensions and physicochemical properties of nanoparticles with unprecedented accuracy and precision. Here, for the
Christopher Polonyi et al.
Dalton transactions (Cambridge, England : 2003), 43(31), 11941-11949 (2014-06-27)
The ability of the anticancer active drug cisplatin to exert biological activity through interference with nucleic acid function is well documented. Since kinetics play a key role in determining product distributions in these systems, methods for accurate documentation of reactivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service