Skip to Content
Merck
All Photos(1)

Key Documents

ERMCZ120

Fine dust (PM10-like) (trace elements)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

Application

  • Seasonal changes and respiratory deposition flux of PM2.5 and PM10 bound metals in Dhaka, Bangladesh: Investigating the seasonal variability of trace elements in PM10-like particles, this research underscores their respiratory impacts in urban settings. It offers a detailed analysis crucial for biochemical researchers focused on the implications of environmental pollution on public health (Moniruzzaman et al., 2022).

  • Mass size distributions, composition and dose estimates of particulate matter in Saharan dust outbreaks: This study evaluates the mass size distributions and composition of PM10 during Saharan dust events, providing essential data for environmental scientists and public health officials. It′s particularly valuable for those studying the transport and deposition of dust-bound trace elements across large geographical areas (Gini et al., 2022).

  • Insights into Elemental Composition and Sources of Fine and Coarse Particulate Matter in Dense Traffic Areas in Toronto and Vancouver, Canada: Analyzing the elemental composition of fine and coarse particulate matter, this research aids in understanding the sources and potential health effects of urban air pollution, emphasizing the role of trace elements in environmental and pharmaceutical pollution studies (Celo et al., 2021).

  • Seasonal variation and sources of carbonaceous species and elements in PM2.5 and PM10 over the eastern Himalaya: This detailed study on the variability of PM10-like fine dust in the eastern Himalaya offers insights into the sources and impacts of particulate matter, supporting researchers engaged in environmental science and pharmacological studies on inhalable particulates (Sharma et al., 2021).

Analysis Note

For more information please see:
ERMCZ120

Legal Information

ERM is a registered trademark of European Commission

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Kyung Seok et al.
Skin pharmacology and physiology, 31(3), 134-143 (2018-03-23)
Airborne particulate matter with a diameter of < 10 µm (PM10) causes oxidative damage, inflammation, and premature skin aging. In this study, we evaluated whether polyphenolic antioxidants attenuate the inflammatory responses of PM10-exposed keratinocytes. Primary human epidermal keratinocytes were exposed
Jung-Won Shin et al.
International journal of molecular sciences, 21(10) (2020-05-18)
Particulate matter (PM), a major air pollutant, is a complex mixture of solid and liquid particles of various sizes. PM has been demonstrated to cause intracellular inflammation in human keratinocytes, and is associated with various skin disorders, including atopic dermatitis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service