Skip to Content
Merck
All Photos(1)

Key Documents

72599

Supelco

Color Reference Solution acc. to APHA

analytical standard, set of 20 × 2 mL 20 standard solutions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

according to ASTM® D 1209-00

shelf life

limited shelf life, expiry date on the label

packaging

set of 20 × 2 mL 20 standard solutions

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

The kit contains a set of 20 color reference solutions defined by ASTM D1209-00, spanning concentrations of platinum cobalt to water from 0 to 500 ppm.
contains Cobalt(II) chloride

Application

The color reference solution finds its use in routine calibration of colour measuring instruments and validation of test results.

Features and Benefits

  • Ready-to-use solutions
  • Compliant to ASTM D 1209-00
  • Available in tamper-evident 2 mL ampules
  • Expiry dated on the label with a limited shelf life
  • Accompanied with a detailed certificate of analysis for an effective quality management system

Packaging

ampules with 2 mL solution

Components

This is a set of the following color reference solutions according to APHA:

  • Colour Reference Solution 5 - 2mL ampule
  • Colour Reference Solution 10 - 2mL ampule
  • Colour Reference Solution 15 - 2mL ampule
  • Colour Reference Solution 20 - 2mL ampule
  • Colour Reference Solution 25 - 2mL ampule
  • Colour Reference Solution 30 - 2mL ampule
  • Colour Reference Solution 35 - 2mL ampule
  • Colour Reference Solution 40 - 2mL ampule
  • Colour Reference Solution 50 - 2mL ampule
  • Colour Reference Solution 60 - 2mL ampule
  • Colour Reference Solution 70 - 2mL ampule
  • Colour Reference Solution 100 - 2mL ampule
  • Colour Reference Solution 150 - 2mL ampule
  • Colour Reference Solution 200 - 2mL ampule
  • Colour Reference Solution 250 - 2mL ampule
  • Colour Reference Solution 300 - 2mL ampule
  • Colour Reference Solution 350 - 2mL ampule
  • Colour Reference Solution 400 - 2mL ampule
  • Colour Reference Solution 450 - 2mL ampule
  • Colour Reference Solution 500 - 2mL ampule

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service