Skip to Content
Merck
All Photos(2)

Key Documents

38166

Supelco

Phenylacetic acid

analytical standard

Synonym(s):

α-Toluic acid, α-Tolylic acid, Benzeneacetic acid, PAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2H
CAS Number:
Molecular Weight:
136.15
Beilstein:
1099647
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

~4 (vs air)

vapor pressure

1 mmHg ( 97 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

265 °C (lit.)

mp

76-78 °C (lit.)

density

1.081 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC(=O)Cc1ccccc1

InChI

1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)

InChI key

WLJVXDMOQOGPHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenylacetic acid may be used as a reference standard in the determination of phenylacetic acid in the methanolic extract of the aerial parts of Justicia wynaadensis using gas-chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GC-MS Analysis of phytocomponents in the methanolic extract of Justicia wynaadensis (nees) T. anders.
Ponnamma US and Manjunath K
International Journal of Pharma and Bio Sciences, 3(3), 570-576 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service