Skip to Content
Merck
All Photos(1)

Key Documents

34244

Supelco

Titanium(IV) oxysulfate solution

1.9-2.1%, for determination of hydrogen peroxyde (H 15), according to DIN 38 409, part 15, DEV-18

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiOSO4
CAS Number:
Molecular Weight:
159.93
MDL number:
UNSPSC Code:
41113319
PubChem Substance ID:
NACRES:
NA.21

quality

for determination of hydrogen peroxyde (H 15), according to DIN 38 409, part 15, DEV-18

Quality Level

specific analyte(s)

peroxidase

concentration

1.9-2.1%

technique(s)

photometry: suitable

impurities

sulfonic acid

suitability

passes test for function test

application(s)

environmental
food and beverages
general analytical

SMILES string

O[Ti+3].O[Ti+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/3H2O4S.2H2O.2Ti/c3*1-5(2,3)4;;;;/h3*(H2,1,2,3,4);2*1H2;;/q;;;;;2*+4/p-8

InChI key

GEXADUANBMINSZ-UHFFFAOYSA-F

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV) oxysulfate is known to form a yellow complex upon reaction with hydrogen peroxide (H2O2) which facilitates the latter′s determination by spectrophotometric methods.

Application

TiOSO4 (titanium (IV) oxysulfate) was used for the preparation of stable amorphous titanium oxide (TiO2) hollow microspheres.
Titanium(IV) oxysulfate solution may be used for the determination of H2O2 in:
  • Municipal wastewater samples by colorimetry and spectrophotometry methods according to DIN 38 409.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integration of Membrane Distillation with solar photo-Fenton for purification of water contaminated with Bacillus sp. and Clostridium sp. Spores.
Ruiz-Aguirre A, et al.
The Science of the Total Environment, 595, 110-118 (2017)
Fabrication of Hollow Inorganic Microspheres by Chemically Induced Self-Transformation.
Yu JG
Advances in Functional Materials, 16 (15), 2035-2041 (2006)
UV/H2O2 process stability and pilot-scale validation for trace organic chemical removal from wastewater treatment plant effluents.
Miklos DB, et al.
Water Research, 136, 169-179 (2018)
Artificial and solar photocatalytic mineralization of psychoactive drugs-loaded urban wastewater: Inorganic ions and phytotoxicity assessment.
Tsoumachidou S, et al.
Process Safety and Environmental Protection, 120, 37-44 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service