Skip to Content
Merck
All Photos(1)

Key Documents

20840

Sigma-Aldrich

Sodium cacodylate trihydrate

purum p.a., ≥98.0% (T)

Synonym(s):

Cacodylic acid sodium salt trihydrate, Dimethylarsinic acid sodium salt, Dimethylarsonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2AsO2Na · 3H2O
CAS Number:
Molecular Weight:
214.03
Beilstein:
3702348
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98.0% (T)
form:
solid
Pricing and availability is not currently available.

grade

purum p.a.

Quality Level

Assay

≥98.0% (T)

form

solid

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤100 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤500 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].[H]O[H].[H]O[H].[H]O[H].C[As](C)([O-])=O

InChI

1S/C2H7AsO2.Na.3H2O/c1-3(2,4)5;;;;/h1-2H3,(H,4,5);;3*1H2/q;+1;;;/p-1

InChI key

RLGWPHBPRCROJO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium cacodylate trihydrate is a biological buffer. Its crystals have been analyzed by single-crystal X-ray diffraction studies. Studies suggested its crystals exhibited two phases, viz. penta-μ-aqua-disodium(I) bis(dimethylarsenate), {[Na2(H2O)5](C2H6AsO2)2}n and di-μ-aqua-bis[triaquasodium(I)] bis(dimethylarsenate), [Na2(H2O)8](C2H6AsO2)2.[1]

Application

Sodium cacodylate trihydrate may be employed as buffer to maintain pH 7.4 in electron microscopic studies.[2][3] It may be used to compose cacodylate buffer for use in electron microscopic studies.[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Zagzag et al.
Cancer research, 62(9), 2660-2668 (2002-05-01)
Enhanced expression of tenascin-C (TN-C) at the invasive edges of glioblastoma multiforme in close association with vascular sprouts, suggests a role for TN-C in microvascular cell migration. To test this hypothesis, we studied the migration of endothelial cells in vitro.
Kathleen A Hinds et al.
Blood, 102(3), 867-872 (2003-04-05)
Tracking transplanted stem cells using magnetic resonance imaging (MRI) could offer biologic insight into homing and engraftment. Ultrasmall dextran-coated iron oxide particles have previously been developed for uptake into cells to allow MRI tracking. We describe a new application of
Anders Lennartson et al.
Acta crystallographica. Section C, Crystal structure communications, 64(Pt 1), m13-m16 (2008-01-25)
Crystals from commercial samples of sodium cacodylate trihydrate, NaO2As(CH3)2 x 3 H2O, were analyzed by single-crystal X-ray diffraction and two phases were identified, viz. penta-mu-aqua-disodium(I) bis(dimethylarsenate), {[Na2(H2O)5](C2H6AsO2)2}n, (I), and di-mu-aqua-bis[triaquasodium(I)] bis(dimethylarsenate), [Na2(H2O)8](C2H6AsO2)2, (II). Both (I) and (II) form layered structures
Aniko Varadi et al.
Journal of cell science, 117(Pt 19), 4389-4400 (2004-08-12)
While the subcellular organisation of mitochondria is likely to influence many aspects of cell physiology, its molecular control is poorly understood. Here, we have investigated the role of the retrograde motor protein complex, dynein-dynactin, in mitochondrial localisation and morphology. Disruption
Z Sobańska et al.
Toxicology in vitro : an international journal published in association with BIBRA, 68, 104931-104931 (2020-07-09)
Significance of MoS2 nanoparticles as a lubricant or drug carriers indicates the need to assess their safety. In the study we analyzed the effects of MoS2 nano- and microparticles and their internalization in vitro, using 2D and 3D culture models

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service