Skip to Content
Merck
All Photos(4)

Key Documents

03143414001

Roche

Genopure Plasmid Midi Kit

kit of for 20 isolations from 5 to 30 ml

Synonym(s):

plasmid isolation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.55

Quality Level

manufacturer/tradename

Roche

packaging

kit of for 20 isolations from 5 to 30 ml

General description

Plasmid recovery was tested with 50 μg purified plasmid. The recovery was >90%, with more than 80% in supercoiled form. The yield of plasmid DNA was determined by isolating pBS from DH5a cells. From 30 mL culture volume with a density of A600 between 3 and 6, >85 μg of plasmid DNA was obtained.

The purity (checked by the ratio of A260/A280) is 1.8 + 0.2.

No RNA is detectable.

The kit components have been tested for the absence of nucleases according to current quality control procedures.

Application

The Genopure Plasmid Midi Kit prepares transfection-grade plasmid DNA in medium quantities (up to 100 μg plasmid) from bacterial cultures. Isolated plasmid is suitable for most molecular biology applications:

  • Transfection
  • Southern blotting
  • Sequencing
  • PCR
  • Restriction analysis
  • Cloning

Features and Benefits

The Genopure Plasmid Midi Kit prepares highly purified plasmid DNA in medium quantities using a modified alkaline lysis method.

  • Save time with ready-to-use reagents.
Purify up to 20 samples (10 minutes hands-on-time/75 minutes overall).
  • Purify all sizes and types of plasmid,
even BAC DNA, since the crude lysate can be filtered to avoid plasmid shearing.
  • Process multiple samples in parallel
using high speed gravity-flow columns.
  • Eliminate the use of hazardous organic compounds
such as cesium chloride, phenol, chloroform, and ethidium bromide.
  • Obtain higher purity plasmid DNA
over plasmid prepared by 2 x cesium chloride gradient centrifugation.

Components

  • Suspension Buffer
  • RNase A
  • Lysis Buffer
  • Neutralization Buffer
  • Equilibration Buffer
  • Wash Buffer
  • Elution Buffer,
  • NucleoBond AX 100 Columns
  • Folded Filters (150 mm diameter)
  • Sealing Rings

Quality

Plasmid DNA purified by this kit has been tested for restriction digestion; pUC 19 was isolated from transformed HB101 as described in the protocol. 1 μg of plasmid was completely digested with 1 U Msp I for 2 hours at +37°C, as shown by agarose gel analysis.

Plasmid recovery was tested with 50 μg purified plasmid. The recovery was >90%, with more than 80% in supercoiled form. The yield of plasmid DNA was determined by isolating pBS from DH5a cells. From 30 ml culture volume with a density of A600 between 3 and 6, >85 μg of plasmid DNA was obtained.

The purity (checked by the ratio of A260/A280) is 1.8 + 0.2.

RNA contamination was analyzed with 3 μg pBS purified with the standard procedure and checked by electrophoresis on an agarose gel. No RNA was detected.

The kit components have been tested for the absence of nucleases according to current Quality Control procedures.

Preparation Note

The isolation procedure is based on a modified alkaline lysis protocol and can be divided into the following steps:The bacteria are partially lysed, allowing the plasmid DNA to escape the cell wall into the supernatant. The larger E. coli chromosomal DNA is trapped in the cell wall. The lysate is cleared of cellular debris by filtration or centrifugation, and the plasmid DNA-containing fraction is loaded onto a pre-equilibrated column. Since the cleared lysate is applied to the column in a low-salt buffer, plasmid DNA binds to the macroporous anion-exchange material in the column. Cellular impurities are eluted from the column with a high-salt wash. Finally, the plasmid DNA is eluted from the column. Recovered DNA is precipitated from the eluate to remove salt and concentrate the plasmid.

Analysis Note

Sample:
E. coli culture that contains a high-copy number plasmid: 5 to 30 mL bacterial culture
E. coli culture that contains a low-copy number plasmid: 10 to 100 mL bacterial culture
Plasmid Size: The isolation procedure is suitable for all sizes of plasmid. Note: Lysates of larger constructs (up to 100 kb) should be cleared by filtration rather than centrifugation to avoid shearing the plasmid.
Time Required: 60 minutes (including filtration of the lysate)
Typical Yield:
High-copy number plasmid: 3 to 5 μg/mL culture
Low-copy number plasmid: 0.2 to 1 μg/mL culture
Product Purity: Isolated plasmid DNA is free of other bacterial components, including RNA.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F

Flash Point(C)

38 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Case of Adaptive Laboratory Evolution (ALE): Biodegradation of Furfural by Pseudomonas pseudoalcaligenes CECT 5344
Ige?o, et al.
Genes, 10, 499-499 (2019)
Fitness effects of blaCTX-M-15-harbouring F2:A1:B? plasmids on their native Escherichia coli ST131 H30Rx hosts
Palkovicova, et al.
The Journal of Antimicrobial Chemotherapy, 77, 2960-2963 (2022)
Ghrelin rapidly elevates protein synthesis in vitro by employing the rpS6K-eEF2K-eEF2 signalling axis
Zhdanov, et al.
Cellular and Molecular Life Sciences, 79 (2022)
Jessica Hoppstädter et al.
Frontiers in immunology, 9, 3111-3111 (2019-02-07)
Activation of toll-like receptors (TLRs) plays a pivotal role in the host defense against bacteria and results in the activation of NF-κB-mediated transcription of proinflammatory mediators. Glucocorticoid-induced leucine zipper (GILZ) is an anti-inflammatory mediator, which inhibits NF-κB activity in macrophages.

Articles

Ion exchange chromatography is employed by the Genopure Plasmid Midi Kit and the Genopure Plasmid Maxi Kit.

Ion exchange chromatography is employed by the Genopure Plasmid Midi Kit and the Genopure Plasmid Maxi Kit.

Ion exchange chromatography is employed by the Genopure Plasmid Midi Kit and the Genopure Plasmid Maxi Kit.

Ion exchange chromatography is employed by the Genopure Plasmid Midi Kit and the Genopure Plasmid Maxi Kit.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service