Skip to Content
Merck
All Photos(1)

Key Documents

MXSMCSP48

Millipore

Milliflex® Pre-filled Media Cassettes

Sabouraud Dextrose Agar, Chloramphenicol, for yeasts, for molds, pack of 48 tests, suitable for bioburden testing

Synonym(s):

Sabouraud Dextrose Agar with Chloramphenicol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160801
NACRES:
NA.74

Agency

EP
USP

Quality Level

sterility

sterile

form

plate (pre-poured agar)

shelf life

6 mo.

feature

ready-to-use

composition

Chloramphenicol

packaging

cassette of 10 mL
pack of 48 tests

manufacturer/tradename

Milliflex®

color

clear light amberslightly opalescent

input

sample type bottled water
sample type cosmetics
sample type beer
sample type pharmaceutical(s)

pH

6.4±0.2 (25.0 °C)

application(s)

bioburden testing
cosmetics
microbiology
pharmaceutical
water monitoring

compatibility

for use with Milliflex®

shipped in

ambient

storage temp.

2-8°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Milliflex® Cassette are ready to use agar-based media to be used with Milliflex® or Milliflex Oasis® system for convenient, rapid and reliable bioburden testing. The prefilled agar cassette helps to streamline the microbial contamination testing workflow with quality assurance and avoiding risks of exogenous contamination. Each plate is a 3-piece assembly containing approximately 10ml of the media. Sabouraud Dextrose Agar is a complex medium for cultivation and isolation of a wide range of yeasts and molds. Dextrose supports the growth of fungi. The low pH favour′s fungal growth and inhibits contaminating bacteria from samples. The medium is also supplemented with chloramphenicol to inhibit the growth of accompanying bacterial flora.

Application

Milliflex® Cassette prefilled with Sabouraud Dextrose Agar is used in Bioburden testing and general microbiological analysis of water especially high purity water (electronics, RO, distilled or purified). It is widely used in pharmaceutical, cosmetic and brewing industry.

Specifications

United-States Pharmacopeia XXII Microbial Limits Tests

Culture Medium

Sabouraud Dextrose w/Chloramphenicol

Analysis Note

QC organisms
C. albicans (ATCC 10231)

S. cerevisiae (ATCC 7754)

E. coli (ATCC 8739) and L. casei (ATCC 393) as negative control
Target Microorganisms: Yeast and Mold
Yeast produce white, creamy colonies. Mold are rough textured and/or filamentous

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service