Skip to Content
Merck
All Photos(1)

Key Documents

MABE2032

Sigma-Aldrich

Anti-MLK1/MRTF-A Antibody, clone 1A11

Synonym(s):

MRTF-A, MKL/myocardin-like protein 1, Megakaryoblastic leukemia 1 protein, Megakaryocytic acute leukemia protein, Myocardin-related transcription factor A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A11, monoclonal

mol wt

calculated mol wt 98.92 kDa
observed mol wt ~140 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG2bκ

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... MRTFA(57591)

Specificity

Clone 1A11 is a mouse monoclonal antibody that detects Myocardin-related transcription factor A (MLK1/MRTF-A). It targets an epitope within the N-terminal region.

Immunogen

GST-tagged recombinant fragment corresponding to 260 amino acids from the N-terminal region of human Myocardin-related transcription factor A (MLK1/MRTF-A).

Application

Quality Control Testing

Evaluated by Western Blotting in Ramos cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MLK1/MRTF-A in Ramos cell lysate.

Tested Applications

Immunofluorescence Analysis: A representative lot detected MLK1/MRTF-A in Immunofluorescence application (Busche, S., et al. (2010). J Cell Sci. 123(Pt16):2803-9).

Western Blotting Analysis: A representative lot detected MLK1/MRTF-A in Western Blotting application (Weissbach, J., et al. (2016). Mol Cell Biol. 36(10):1526-39).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Myocardin-related transcription factor A (UniProt: Q969V6; also known as MRTF-A, MKL/myocardin-like protein 1, Megakaryoblastic leukemia 1 protein, Megakaryocytic acute leukemia protein) is encoded by the MRTFA (also known as KIAA1438, MAL, MLK1) gene (Gene ID: 57591) in human. MRTF-A is a ubiquitously expressed transcription coactivator that associates with serum response factor (SRF) transcription factor to control expression of genes that regulate cytoskeleton during development, morphogenesis, and cell migration. The MRTFA-SRF complex activity responds to Rho GTPase-induced changes in cellular G-actin concentration, and thereby couples cytoskeletal gene expression to cytoskeletal dynamics. It is reported to bind to G-actin via its RPEL repeats. It is detected in cytoplasm as well as in the nucleus and its subcellular location is tightly regulated by actin. High levels of G-actin in the nucleus, observed during serum deprivation, result in low levels of nuclear MRTF-A, while reduced levels of nuclear G-actin result in accumulation of MRTF-A in the nucleus. G-actin-binding in the cytoplasm inhibits its nuclear import by masking the nuclear localization signal (NLS). In contrast, binding to nuclear G-actin promotes its nuclear export to the cytoplasm. Phosphorylation of serine 6 by ERK inhibits the binding of G-actin, unmasking the NLS and promoting nuclear import. Its N-terminal region is required for its nuclear localization and the C-terminal region mediates its transcriptional activity. (Ref.: Weissbach, J., et al. (2016). Mol. Cell. Biol. 36(10); 1526-1539; Busche, S., et al. (2010). J. Cell Sci. 123(16); 2803-2809).

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service