Skip to Content
Merck
All Photos(1)

Key Documents

MAB3317

Sigma-Aldrich

Anti-MMP-14 Antibody, hemopexin domain, clone 113-5B7

clone 113-5B7, Chemicon®, from mouse

Synonym(s):

MT1-MMP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

113-5B7, monoclonal

species reactivity

rat, human, bovine, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG3κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MMP14(4323)

Specificity

Reacts with human MMP-14, also known as MT1-MMP, with a very slight cross-reactivity to hMMP-3. For unknown reasons this antibody does not react on westerns to CC1043.

Immunogen

CDGNFDTVAMLRGEM in the hemopexin-like domain of human MT1-MMP
Epitope: a.a. 319-333

Application

Anti-MMP-14 Antibody, hemopexin domain, clone 113-5B7 is an antibody against MMP-14 for use in ELISA, IH(P) & WB.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
Western Blot: 10 μg/mL, membrane preparations; MD-MB-231 cells pretreated with 1-5 μg/mL ConA for 24-48 hours as positive control. 60-65 kDa proteins are identified.


When tested against recombinant MT1-MMP (lacking the TM domain) expressed in CHO cells the antibody reacts with a band of ~59 kDa.


Immunohistochemistry: Frozen tissue sections; traditional formalin fixation is problematic. Paraffin-embedded tissue sections with PLP fixation required.


Immunoprecipitation


EIA


Optimal working dilutions must be determined by the end user.

Target description

~59-69 kDa

Physical form

Format: Purified
Protein A purified
Purified immunoglobulin. Liquid in 0.1 M sodium phosphate buffer, pH 7.0, containing 2% protease-free bovine Serum albumin.

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Breast carcinoma tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Manufactured by Daiichi Fine Chemical Co., Ltd

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic expression of matrix metalloproteinases (MMP-2, -9 and -14) and the tissue inhibitors of MMPs (TIMP-1, -2 and -3) at the implantation site during tubal pregnancy.
Bai, SX; Wang, YL; Qin, L; Xiao, ZJ; Herva, R; Piao, YS
Reproduction (Cambridge, England) null
Jeffrey J Atkinson et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 232(4), 1079-1090 (2005-03-02)
Matrix metalloproteinases (MMPs) are expressed during lung development, but their role may be limited, as mice deficient in MMP-3, 7, 9, or 12 develop a normal adult lung. Because membrane-type 1 matrix metalloproteinase (MT1-MMP) is expressed in the developing lung
Riccardo E Nisato et al.
Cancer research, 65(20), 9377-9387 (2005-10-19)
Matrix metalloproteinase (MMP)-2 and its hemopexin C domain autolytic fragment (also called PEX) have been proposed to be crucial for angiogenesis. Here, we have investigated the dependency of in vitro angiogenesis on MMP-mediated extracellular proteolysis and integrin alpha(v)beta3-mediated cell adhesion
Amit Ranjan et al.
BioMed research international, 2014, 804680-804680 (2014-09-03)
Matrix remodeling and invasion of basement membrane are the major determinants of malignant progression. Matrix degrading enzymes play a pivotal role in this process and have been shown to be regulated at multiple levels. Using high metastatic B16F10 and its
Nor E Sounni et al.
Disease models & mechanisms, 3(5-6), 317-332 (2010-03-13)
Innate regulatory networks within organs maintain tissue homeostasis and facilitate rapid responses to damage. We identified a novel pathway regulating vessel stability in tissues that involves matrix metalloproteinase 14 (MMP14) and transforming growth factor beta 1 (TGFbeta(1)). Whereas plasma proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service