Skip to Content
Merck
All Photos(1)

Key Documents

AB5374

Sigma-Aldrich

Anti-Potassium Channel Kir2.1 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNJ2(3759)

Specificity

Recognizes Kir2.1 protein (IRK1, Kcnj2). The epitope is specific for Kir2.1 and is not present in any other known protein.

SPECIES REACTIVITIES: The immunogen sequence is identical in rabbit, bovine, porcine and guinea pig. It is highly conserved in rat and mouse (17/19 residues), chick (15/19 residues), and pigeon (14/18). Other species have not been tested.

Immunogen

Purified peptide from human Kir2.1 (amino acids 392-410) (Accession number P48049).

Application

Detect Potassium Channel Kir2.1 using this Anti-Potassium Channel Kir2.1 Antibody validated for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1:200 using ECL on rat brain and rat heart membranes. Note: Addition of 0.1% Tween to the standard milk block is recommended for this lot.

Immunohistochemistry on rat heart sections.

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 20 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Kristensen et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 290(3), R766-R772 (2005-10-15)
Muscle activity is associated with potassium displacements, which may cause fatigue. It was reported previously that the density of the large-conductance Ca2+-dependent K+ (BK(Ca)) channel is higher in the T tubule membrane than in the sarcolemmal membrane and that the
Isabelle Deschênes et al.
Journal of molecular and cellular cardiology, 45(3), 336-346 (2008-06-21)
The Ca(2+)-independent transient outward potassium current (I(to)) encoded by the Kv4 family of potassium channels, is central to normal repolarization of cardiac myocytes. KChIPs are a group of Ca(2+)-binding accessory subunits that modulate Kv4-encoded currents. However, the biophysical effects of
Fadi G Akar et al.
American journal of physiology. Heart and circulatory physiology, 288(6), H2887-H2896 (2005-02-01)
Heart failure (HF) is characterized by marked prolongation of action potential duration and reduction in cellular repolarization reserve. These changes are caused in large part by HF-induced K(+) current downregulation. Molecular mechanisms underlying these changes remain unclear. We determined whether
Lisa Chilton et al.
Journal of the American Society of Nephrology : JASN, 19(1), 69-76 (2008-01-08)
The afferent and efferent arterioles regulate the inflow and outflow resistance of the glomerulus, acting in concert to control the glomerular capillary pressure and glomerular filtration rate. The myocytes of these two vessels are remarkably different, especially regarding electromechanical coupling.
Alejandro Vila et al.
The Journal of comparative neurology, 525(4), 850-867 (2016-08-26)
Synaptic processes and plasticity of synapses are mediated by large suites of proteins. In most cases, many of these proteins are tethered together by synaptic scaffold proteins. Scaffold proteins have a large number and typically a variety of protein interaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service