Skip to Content
Merck
All Photos(1)

Key Documents

8.22029

Sigma-Aldrich

3,4-Dihydroxycinnamic acid

for synthesis

Synonym(s):

3,4-Dihydroxycinnamic acid, Caffeic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(OH)2C6H3CHCHCOOH
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
12352106
EC Index Number:
206-361-2
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

form

powder

mp

234-237 °C (decomposition)

storage temp.

2-30°C

SMILES string

Oc1c(ccc(c1)\C=C/C(=O)O)O

InChI

1S/C9H8O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1-5,10-11H,(H,12,13)/b4-2-

InChI key

QAIPRVGONGVQAS-RQOWECAXSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Why are the melting points of caffeic acid 8220290010 (listed as 3,4-dihydroxycinnamic acid) and caffeic acid C0625 so different? They are both listed as >98% (HPLC).

    1 answer
    1. The determination of melting point largely depends on testing conditions, such as equipment type, the rate of temperature increase, the diameter of the glass tube, etc. These products are not tested for melting point. These products are from different brands and melting point values are taken from different literature sources.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service