Skip to Content
Merck
All Photos(1)

Key Documents

8.08671

Sigma-Aldrich

2,6-Dimethylaniline

for synthesis

Synonym(s):

2,6-Dimethylaniline, vic.-m-Xylidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,6-(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
MDL number:
UNSPSC Code:
12352116
EC Index Number:
201-758-7
NACRES:
NA.22

vapor pressure

0.2 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

490 °C

potency

1290 mg/kg LD50, oral (Rat)

expl. lim.

1.3-6.9 % (v/v)

pH

12.5 (20 °C, 100 g/L in H2O)

bp

216 °C/1013 hPa

mp

10-12 °C

transition temp

flash point 92 °C

solubility

13 g/L

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5H,9H2,1-2H3

InChI key

UFFBMTHBGFGIHF-UHFFFAOYSA-N

Application

2,6-Dimethylaniline for synthesis. CAS 87-62-7, chemical formula 2,6-(CH₃)₂C₆H₃NH₂.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.978 - 0.981
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service